missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Bromo-10-(1-naphthyl)anthracene, 98%

Manufacturer:  Alfa Aesar™ H6402406

 View more versions of this product

Catalog No. AAH6402406



Specifications

Specifications

9-Bromo-10-(1-naphthyl)anthracene
5g
C24H15Br
20386637
SYACRXBYRNYMLN-UHFFFAOYSA-N
9-bromo-10-naphthalen-1-ylanthracene
21076365
Solid
Yellow
400607-04-7
MFCD11046571
9-bromo-10-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl anthracene, 9-bromo-10-1-naphthyl anthracene, anthracene, 9-bromo-10-1-naphthalenyl, anthracene, 9-bromo-10-1-naphthalenyl, 9-bromo-10-naphthalen-1-yl-anthracene, 10-bromo-9-phthalen-1-yl anthracene, 10-bromo-9-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl-anthracene, 10-naphthalene-1-yl-9-bromo anthracene
C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br
383.288
383.29
98%

Chemical Identifiers

400607-04-7
383.288
SYACRXBYRNYMLN-UHFFFAOYSA-N
21076365
C1=CC=C2C(=C1)C=CC=C2C3=C4C=CC=CC4=C(C5=CC=CC=C53)Br
C24H15Br
MFCD11046571
9-bromo-10-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl anthracene, 9-bromo-10-1-naphthyl anthracene, anthracene, 9-bromo-10-1-naphthalenyl, anthracene, 9-bromo-10-1-naphthalenyl, 9-bromo-10-naphthalen-1-yl-anthracene, 10-bromo-9-phthalen-1-yl anthracene, 10-bromo-9-naphthalen-1-yl anthracene, 9-bromo-10-1-naphthalenyl-anthracene, 10-naphthalene-1-yl-9-bromo anthracene
9-bromo-10-naphthalen-1-ylanthracene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents