missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-beta-D-Arabinofuranosyl-2-fluoroadenine-5'-monophosphate, 99%

$53.66 - $192.77

Chemical Identifiers

CAS 75607-67-9
Molecular Formula C10H13FN5O7P
Molecular Weight (g/mol) 365.214
MDL Number MFCD00866418
InChI Key GIUYCYHIANZCFB-FJFJXFQQSA-N
Synonym fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine phosphate, fludarabine monophosphate, famp, unii-1x9vk9o1sc
PubChem CID 30751
ChEBI CHEBI:63599
IUPAC Name [(2R,3S,4S,5R)-5-(6-amino-2-fluoropurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)N=C(N=C2N)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65154MD View Documents Alfa Aesar™
J65154MD
250mg Each for $53.66
 
AAJ6515403 View Documents Alfa Aesar™
J6515403
1g Each for $192.77
 
Description

Description

Solubility
Soluble in DMSO or water at 5mg/ml

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Specifications

Chemical Identifiers

75607-67-9
365.214
GIUYCYHIANZCFB-FJFJXFQQSA-N
30751
[(2R,3S,4S,5R)-5-(6-amino-2-fluoropurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
C10H13FN5O7P
MFCD00866418
fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine phosphate, fludarabine monophosphate, famp, unii-1x9vk9o1sc
CHEBI:63599
C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)N=C(N=C2N)F

Specifications

White
75607-67-9
MFCD00866418
Soluble in DMSO or water at 5mg/ml
C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)N=C(N=C2N)F
365.214
CHEBI:63599
Powder
9-beta-D-Arabinofuranosyl-2-fluoroadenine-5'-monophosphate
250mg
C10H13FN5O7P
fludarabine phosphate, fludara, fludarabine 5'-monophosphate, fludura, oforta, 2-fluoro-ara amp, fludarabine phosphate, fludarabine monophosphate, famp, unii-1x9vk9o1sc
GIUYCYHIANZCFB-FJFJXFQQSA-N
[(2R,3S,4S,5R)-5-(6-amino-2-fluoropurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
30751
365.21
99%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UO7440900

Recommended Storage : 2° to 8°C