missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Benzylcarbazole-3-carboxaldehyde, TCI America™

$207.78 - $656.95

Chemical Identifiers

CAS 54117-37-2
Molecular Formula C20H15NO
Molecular Weight (g/mol) 285.346
InChI Key GSNXZYWQXATWRX-UHFFFAOYSA-N
Synonym 9-Benzyl-3-formylcarbazole
PubChem CID 3091534
IUPAC Name 9-benzylcarbazole-3-carbaldehyde
SMILES C1=CC=C(C=C1)CN2C3=C(C=C(C=C3)C=O)C4=CC=CC=C42
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28041G View Documents TCI America
B28041G
1g Each for $207.78
 
B28045G View Documents TCI America
B28045G
5g Each for $656.95
 
Specifications

Chemical Identifiers

54117-37-2
285.346
9-Benzyl-3-formylcarbazole
9-benzylcarbazole-3-carbaldehyde
C20H15NO
GSNXZYWQXATWRX-UHFFFAOYSA-N
3091534
C1=CC=C(C=C1)CN2C3=C(C=C(C=C3)C=O)C4=CC=CC=C42

Specifications

Undesignated
1g
C20H15NO
GSNXZYWQXATWRX-UHFFFAOYSA-N
9-benzylcarbazole-3-carbaldehyde
3091534
Crystal-Powder at 20°C
9-Benzylcarbazole-3-carboxaldehyde
134°C
54117-37-2
9-Benzyl-3-formylcarbazole
C1=CC=C(C=C1)CN2C3=C(C=C(C=C3)C=O)C4=CC=CC=C42
285.346
285.35
≥97.0% (N)
Safety and Handling

Safety and Handling

TSCA : No