Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Benzylcarbazole-3-boronic acid pinacol ester, 95%

$68.26 - $275.74

Chemical Identifiers

Chemical Name or Material 9-Benzylcarbazole-3-boronic acid pinacol ester
Molecular Formula C25H26BNO2
Formula Weight 383.3
MDL Number MFCD16294547
Synonym 9-Benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5194403

  SDS
alfa aesar™
H5194403
1g Each for $68.26

AAH5194406

  SDS
alfa aesar™
H5194406
5g Each for $275.74
Description & Specifications

Specifications

Formula Weight 383.3
MDL Number MFCD16294547
Molecular Formula C25H26BNO2
Assay 95%
Chemical Name or Material 9-Benzylcarbazole-3-boronic acid pinacol ester
Synonym 9-Benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Quantity 1g