Please login to your online account to display your discounted pricing

9-Benzhydrylidene-10-anthrone, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC388260010

 View more versions of this product

Catalog No. AC388260010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 358.44
Quantity 1g
Color Beige-yellow
EINECS Number 211-570-7
Melting Point 200° to 204°C
Beilstein 07, 551
CAS Max % 100.0
CAS Min % 94.0
Assay Percent Range 98%
Molecular Formula C27H18O
MDL Number MFCD00019172
Packaging Glass bottle
Chemical Name or Material 9-Benzhydrylidene-10-anthrone
CAS 667-91-4
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 9-Benzhydrylidene-10-anthrone
CAS 667-91-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 98%
Molecular Formula C27H18O
Formula Weight 358.44
MDL Number MFCD00019172