missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-Anthracenemethanol, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC181872500

 View more versions of this product

Catalog No. AC181872500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

9-Anthracenemethanol
Glass bottle
160.0°C to 164.0°C
98%
97.5
98%
MFCD00001264
JCJNNHDZTLRSGN-UHFFFAOYSA-N
anthracen-9-ylmethanol
73848
Crystalline Powder
Authentic
Yellow
250g
1468-95-7
100.0
C15H12O
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
208.26
208.26
98%

Chemical Identifiers

1468-95-7
208.26
JCJNNHDZTLRSGN-UHFFFAOYSA-N
73848
C1=CC=C2C(=C1)C=C3C=CC=CC3=C2CO
C15H12O
MFCD00001264
9-anthracenemethanol, 9-hydroxymethylanthracene, 9-anthrylcarbinol, 9-methylolanthracene, 9-anthrylmethanol, 9-hydroxymethylantracene, 9-anthracene methanol, 9-hydroxymethyl anthracene, anthracene-9-carbinol, ccris 7297
anthracen-9-ylmethanol
Safety and Handling

Safety and Handling

EINECSNumber : 215-998-5

RTECSNumber : CB0577500

TSCA : TSCA

SDS Documents