Learn More
9-Aminophenanthrene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H55773MD
Description
9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9-Aminophenanthrene can be used as a model ligand to study the binding of 9-aminophenanthrene to individual subsites within the active site of CYP(eryF). It is also used as a fluorescent-labeling reagent for free fatty acids (FFA),examination through HPLC.
Solubility
It is slightly soluble in water 0.00581 mg/mL.
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
9-Aminophenanthrene | |
137°C to 139°C | |
C14H11N | |
250 mg | |
It is slightly soluble in water 0.00581mg/mL. | |
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)N | |
193.249 | |
CHEBI:50475 | |
96% |
947-73-9 | |
White to Brown | |
MFCD00001177 | |
9-aminophenanthrene, 9-phenanthrenamine, 9-phenanthrylamine, phenanthren-9-ylamine, unii-7m9m5km2xy, ccris 7006, 7m9m5km2xy, 9ap, acmc-20alvr, phenanthren-9-yl-amine | |
KIHQWOBUUIPWAN-UHFFFAOYSA-N | |
phenanthren-9-amine | |
13695 | |
193.24 | |
Solid |
Chemical Identifiers
947-73-9 | |
193.249 | |
KIHQWOBUUIPWAN-UHFFFAOYSA-N | |
13695 | |
phenanthren-9-amine |
C14H11N | |
MFCD00001177 | |
9-aminophenanthrene, 9-phenanthrenamine, 9-phenanthrylamine, phenanthren-9-ylamine, unii-7m9m5km2xy, ccris 7006, 7m9m5km2xy, 9ap, acmc-20alvr, phenanthren-9-yl-amine | |
CHEBI:50475 | |
C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)N |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-431-6
RTECSNumber : SG0175000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only