Please login to your online account to display your discounted pricing

Alfa Aesar™ 9-Aminoacridine hydrochloride hydrate, 99%

Chemical Identifiers

Chemical Name or Material 9-Aminoacridine hydrochloride hydrate
CAS 52417-22-8
Molecular Formula C13H10N2·HCl·xH2O
Formula Weight 230.69 (Anhydrous)
MDL Number MFCD00150071
UN Number UN2811
Synonym 9-Acridinamine hydrochloride hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2435606

  SDS
alfa aesar™
B2435606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2435614

  SDS
alfa aesar™
B2435614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.69 (Anhydrous)
Chemical Name or Material 9-Aminoacridine hydrochloride hydrate
TSCA TSCA
Assay 99%
Molecular Formula C13H10N2·HCl·xH2O
MDL Number MFCD00150071
CAS 52417-22-8
UN Number UN2811
Melting Point >300°C
Synonym 9-Acridinamine hydrochloride hydrate
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H341
Toxic if swallowed.
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number AR7350000

Applications
9-Aminoacridine hydrochloride is a known mutagen and DNA modifier.

Solubility
Soluble in water (3.3 g/L) at 20°C.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H341
Toxic if swallowed.
Suspected of causing genetic defects.