missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 97.0+%, TCI America™
$217.33 - $733.29
Chemical Identifiers
CAS | 958261-51-3 |
---|---|
Molecular Formula | C41H65B2NO4 |
Molecular Weight (g/mol) | 657.594 |
MDL Number | MFCD16621134 |
InChI Key | XMKFCPVHTTWWCK-UHFFFAOYSA-N |
Synonym | 9-(1-n-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, 9-(1-n-Octylnonyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(9-Heptadecanyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester |
PubChem CID | 51341870 |
IUPAC Name | 9-heptadecan-9-yl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C(CCCCCCCC)CCCCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
O0428200MG
|
TCI America
O0428200MG |
200 mg |
Each for $217.33
|
|
|||||
O04281G
|
TCI America
O04281G |
1 g |
Each for $733.29
|
|
|||||
Chemical Identifiers
958261-51-3 | |
657.594 | |
XMKFCPVHTTWWCK-UHFFFAOYSA-N | |
51341870 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C(CCCCCCCC)CCCCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C |
C41H65B2NO4 | |
MFCD16621134 | |
9-(1-n-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, 9-(1-n-Octylnonyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(9-Heptadecanyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester | |
9-heptadecan-9-yl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole |
Specifications
958261-51-3 | |
White | |
MFCD16621134 | |
9-(1-n-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, 9-(1-n-Octylnonyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(9-Heptadecanyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C(CCCCCCCC)CCCCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C | |
657.594 | |
657.59 | |
Crystalline Powder |
146°C | |
C41H65B2NO4 | |
200 mg | |
XMKFCPVHTTWWCK-UHFFFAOYSA-N | |
9-heptadecan-9-yl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole | |
51341870 | |
≥97.0% (HPLC,N) | |
9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole |
Safety and Handling
TSCA : No