Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,9'-Dixanthylidene, 98%

$334.50 - $334.50

Chemical Identifiers

Chemical Name or Material 9,9'-Dixanthylidene
CAS 517-45-3
Molecular Formula C26H16O2
Formula Weight 360.41
MDL Number MFCD00597968
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2173006

  SDS
alfa aesar™
B2173006
5g Each for $334.50
Description & Specifications

Specifications

Formula Weight 360.41
Assay 98%
CAS 517-45-3
Melting Point 318° to 320°C
Quantity 5g
Chemical Name or Material 9,9'-Dixanthylidene
Molecular Formula C26H16O2
MDL Number MFCD00597968