Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,9-Dimethylxanthene, 98+%

$43.41 - $135.15

Chemical Identifiers

Chemical Name or Material 9,9-Dimethylxanthene
CAS 19814-75-6
Molecular Formula C15H14O
Formula Weight 210.28
MDL Number MFCD00134434
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2734003

  SDS
alfa aesar™
H2734003
1g Each for $43.41

AAH2734006

  SDS
alfa aesar™
H2734006
5g Each for $135.15
Description & Specifications

Specifications

Formula Weight 210.28
Chemical Name or Material 9,9-Dimethylxanthene
Melting Point 40° to 43°C
Boiling Point 114° to 115°C (0.6mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Assay ≥98%
Molecular Formula C15H14O
MDL Number MFCD00134434
CAS 19814-75-6

Applications
9,9-Dimethylxanthene is suitable for use in the preparation of 9,9-dimethylxanthene-4,5-dicarboxylic acid. It may be used in the preparation of 9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene. It may be used in the preparation of 4,6-bis(dimesitylboryl)dibenzofuran.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.