missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9-Dimethylfluorene 98.0+%, TCI America™
$124.89 - $365.35
Chemical Identifiers
| CAS | 4569-45-3 |
|---|---|
| Molecular Formula | C15H14 |
| Molecular Weight (g/mol) | 194.28 |
| MDL Number | MFCD00114670,MFCD31699973 |
| InChI Key | ZHQNDEHZACHHTA-UHFFFAOYSA-N |
| PubChem CID | 78325 |
| IUPAC Name | 9,9-dimethyl-9H-fluorene |
| SMILES | CC1(C)C2=CC=CC=C2C2=CC=CC=C12 |
Chemical Identifiers
| 4569-45-3 | |
| 194.28 | |
| ZHQNDEHZACHHTA-UHFFFAOYSA-N | |
| 9,9-dimethyl-9H-fluorene |
| C15H14 | |
| MFCD00114670,MFCD31699973 | |
| 78325 | |
| CC1(C)C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
| 4569-45-3 | |
| White | |
| C15H14 | |
| 1 g | |
| CC1(C)C2=CC=CC=C2C2=CC=CC=C12 | |
| 194.28 | |
| 194.28 | |
| Crystalline Powder |
| 97°C | |
| 287°C | |
| MFCD00114670,MFCD31699973 | |
| ZHQNDEHZACHHTA-UHFFFAOYSA-N | |
| 9,9-dimethyl-9H-fluorene | |
| 78325 | |
| ≥98.0% (GC) | |
| 9,9-Dimethylfluorene |
Safety and Handling
TSCA : No