missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9,9-Dimethylfluorene-2-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5195006
Description
It is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in the synthesis and photo physical properties of 1,2-diphenylindolizine derivatives: fluorescent blue-emitting materials for organic light-emitting device.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
9,9-Dimethylfluorene-2-boronic acid pinacol ester | |
C21H25BO2 | |
5 g | |
DAZFRJAIIUPRQZ-UHFFFAOYSA-N | |
2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
45358694 | |
95% |
569343-09-5 | |
MFCD08704229 | |
2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2-boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl, 2-9,9-dimethylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fl-be, amtb252, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9,9-dimethyl-9h-fluorene | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(C)C | |
320.239 | |
320.24 |
Chemical Identifiers
569343-09-5 | |
320.239 | |
DAZFRJAIIUPRQZ-UHFFFAOYSA-N | |
45358694 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(C)C |
C21H25BO2 | |
MFCD08704229 | |
2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2-boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl, 2-9,9-dimethylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fl-be, amtb252, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9,9-dimethyl-9h-fluorene | |
2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only