missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,9-Dimethylfluorene-2-boronic acid, 98%

$24.05 - $257.54

Chemical Identifiers

CAS 333432-28-3
Molecular Formula C15H15BO2
Molecular Weight (g/mol) 238.093
MDL Number MFCD08704227
InChI Key DMDPAJOXRYGXCB-UHFFFAOYSA-N
Synonym 9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-boronic acid, 9,9-dimethylfluoren-2-boronic acid, 9,9-dimethyl-2-fluoreneboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid, 9,9-dimethylfluorene-2-boronic acid, 9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluorene-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronicacid
PubChem CID 22348454
IUPAC Name (9,9-dimethylfluoren-2-yl)boronic acid
SMILES B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64777MD View Documents Alfa Aesar™
H64777MD
250mg Each for $24.05
 
AAH6477703 View Documents Alfa Aesar™
H6477703
1g Each for $68.93
 
AAH6477706 View Documents Alfa Aesar™
H6477706
5g Each for $257.54
 
Specifications

Chemical Identifiers

333432-28-3
238.093
DMDPAJOXRYGXCB-UHFFFAOYSA-N
22348454
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
C15H15BO2
MFCD08704227
9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-boronic acid, 9,9-dimethylfluoren-2-boronic acid, 9,9-dimethyl-2-fluoreneboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid, 9,9-dimethylfluorene-2-boronic acid, 9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluorene-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronicacid
(9,9-dimethylfluoren-2-yl)boronic acid

Specifications

250mg
C15H15BO2
9563230
DMDPAJOXRYGXCB-UHFFFAOYSA-N
(9,9-dimethylfluoren-2-yl)boronic acid
22348454
Solid
9,9-Dimethylfluorene-2-boronic acid
333432-28-3
MFCD08704227
9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-boronic acid, 9,9-dimethylfluoren-2-boronic acid, 9,9-dimethyl-2-fluoreneboronic acid, 9,9-dimethyl-9h-fluoren-2-yl-2-boronic acid, 9,9-dimethylfluorene-2-boronic acid, 9,9-dimethyl-9h-fluoren-2-yl boronic acid, 9,9-dimethyl-9h-fluorene-2-ylboronic acid, 9,9-dimethyl-9h-fluoren-2-ylboronicacid
B(C1=CC2=C(C=C1)C3=CC=CC=C3C2(C)C)(O)O
238.093
238.09
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C