Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester, 97%

Chemical Identifiers

Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester
CAS 317802-08-7
Formula Weight 558.42
MDL Number MFCD03701607
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5539203

 
alfa aesar™
H5539203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55392MD

 
alfa aesar™
H55392MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 558.42
CAS 317802-08-7
Quantity 1g
Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester
MDL Number MFCD03701607
Physical Form Solid
Assay 97%