missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H55392MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester | |
C35H52B2O4 | |
250 mg | |
KAYXDWIILRESPY-UHFFFAOYSA-N | |
2-[7-(1,3,2-dioxaborinan-2-yl)-9,9-dioctylfluoren-2-yl]-1,3,2-dioxaborinane | |
4378560 | |
97% |
317802-08-7 | |
MFCD03701607 | |
9,9-dioctylfluorene-2,7-diboronic acid bis 1,3-propanediol ester, 1,3,2-dioxaborinane, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethylborate, 9 9-dioctylfluorene-2 7-bis trimethylbo, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctylfluoren-2-yl-1,3,2-dioxaborinane, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctyl-fluoren-2-yl-1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethyleneborate, 2,2-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane | |
CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B2OCCCO2)C2=C1C=C(C=C2)B1OCCCO1 | |
558.42 | |
558.42 | |
Solid |
Chemical Identifiers
317802-08-7 | |
558.42 | |
KAYXDWIILRESPY-UHFFFAOYSA-N | |
4378560 | |
CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(=C2)B2OCCCO2)C2=C1C=C(C=C2)B1OCCCO1 |
C35H52B2O4 | |
MFCD03701607 | |
9,9-dioctylfluorene-2,7-diboronic acid bis 1,3-propanediol ester, 1,3,2-dioxaborinane, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, 2,2'-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethylborate, 9 9-dioctylfluorene-2 7-bis trimethylbo, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctylfluoren-2-yl-1,3,2-dioxaborinane, 2-7-1,3,2-dioxaborinan-2-yl-9,9-dioctyl-fluoren-2-yl-1,3,2-dioxaborinane, 9,9-dioctylfluorene-2,7-bis trimethyleneborate, 2,2-9,9-dioctyl-9h-fluorene-2,7-diyl bis 1,3,2-dioxaborinane | |
2-[7-(1,3,2-dioxaborinan-2-yl)-9,9-dioctylfluoren-2-yl]-1,3,2-dioxaborinane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only