missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5537303

 View more versions of this product

Catalog No. AAH5537303



Specifications

Specifications

9,9-Di-n-octylfluorene-2,7-diboronic acid
258865-48-4
MFCD03701608
HURJMQMZDPOUOU-UHFFFAOYSA-N
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid
4283240
Solid
1g
C29H44B2O4
7-borono-9,9-dioctyl-2-fluorenyl boronic acid
B(C1=CC2=C(C=C1)C3=C(C2(CCCCCCCC)CCCCCCCC)C=C(C=C3)B(O)O)(O)O
478.287
478.29
97%

Chemical Identifiers

258865-48-4
478.287
HURJMQMZDPOUOU-UHFFFAOYSA-N
4283240
B(C1=CC2=C(C=C1)C3=C(C2(CCCCCCCC)CCCCCCCC)C=C(C=C3)B(O)O)(O)O
C29H44B2O4
MFCD03701608
7-borono-9,9-dioctyl-2-fluorenyl boronic acid
(7-borono-9,9-dioctylfluoren-2-yl)boronic acid
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents