missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9,9-Di-n-hexylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5581703
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
9,9-Di-n-hexylfluorene-2,7-diboronic acid | |
C25H36B2O4 | |
1 g | |
QFWOJNOZWMHNTK-UHFFFAOYSA-N | |
[7-(dihydroxyboranyl)-9,9-dihexyl-9H-fluoren-2-yl]boronic acid | |
4524285 | |
97% |
203927-98-4 | |
MFCD03427279 | |
9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene | |
CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O | |
422.18 | |
422.18 | |
Solid |
Chemical Identifiers
203927-98-4 | |
422.18 | |
QFWOJNOZWMHNTK-UHFFFAOYSA-N | |
4524285 |
C25H36B2O4 | |
MFCD03427279 | |
9,9-dihexylfluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid, 7-borono-9,9-dihexyl-fluoren-2-yl boronic acid, boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis, acmc-20alvw, 9,9-dihexyl-fluorene-2,7-diboronic acid, 9,9-dihexyl-9h-fluorene-2,7-diboronic acid, 7-borono-9,9-dihexylfluoren-2-yl boronic acid, 2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene | |
CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only