missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene 97.0+%, TCI America™
$127.75 - $441.30
Chemical Identifiers
CAS | 510775-24-3 |
---|---|
Molecular Formula | C57H40N2 |
Molecular Weight (g/mol) | 752.961 |
InChI Key | LKFCORSLOUZJMB-UHFFFAOYSA-N |
PubChem CID | 60106020 |
IUPAC Name | N-[4-[9-[4-(N-naphthalen-1-ylanilino)phenyl]fluoren-9-yl]phenyl]-N-phenylnaphthalen-1-amine |
SMILES | C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3(C4=CC=CC=C4C5=CC=CC=C53)C6=CC=C(C=C6)N(C7=CC=CC=C7)C8=CC=CC9=CC=CC=C98)C1=CC=CC2=CC=CC=C21 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B4979200MG
|
TCI America
B4979200MG |
200 mg |
Each for $127.75
|
|
|||||
B49791G
|
TCI America
B49791G |
1 g |
Each for $441.30
|
|
|||||
Chemical Identifiers
510775-24-3 | |
752.961 | |
60106020 | |
C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3(C4=CC=CC=C4C5=CC=CC=C53)C6=CC=C(C=C6)N(C7=CC=CC=C7)C8=CC=CC9=CC=CC=C98)C1=CC=CC2=CC=CC=C21 |
C57H40N2 | |
LKFCORSLOUZJMB-UHFFFAOYSA-N | |
N-[4-[9-[4-(N-naphthalen-1-ylanilino)phenyl]fluoren-9-yl]phenyl]-N-phenylnaphthalen-1-amine |
Specifications
510775-24-3 | |
C57H40N2 | |
LKFCORSLOUZJMB-UHFFFAOYSA-N | |
N-[4-[9-[4-(N-naphthalen-1-ylanilino)phenyl]fluoren-9-yl]phenyl]-N-phenylnaphthalen-1-amine | |
60106020 | |
≥97.0% (HPLC,N) | |
9,9-Bis[4-[N-(1-naphthyl)anilino]phenyl]fluorene |
White-Yellow | |
200 mg | |
C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3(C4=CC=CC=C4C5=CC=CC=C53)C6=CC=C(C=C6)N(C7=CC=CC=C7)C8=CC=CC9=CC=CC=C98)C1=CC=CC2=CC=CC=C21 | |
752.961 | |
752.96 | |
Crystalline Powder |
Safety and Handling
TSCA : No