missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9,9-Bis(4-methoxyphenyl)-9H-fluorene, TCI America™

$249.03 - $747.09

Chemical Identifiers

CAS 117766-40-2
Molecular Formula C27H22O2
Molecular Weight (g/mol) 378.471
InChI Key BYDIWLYAWBNCAQ-UHFFFAOYSA-N
PubChem CID 11783778
IUPAC Name 9,9-bis(4-methoxyphenyl)fluorene
SMILES COC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B56175G View Documents TCI America
B56175G
5g Each for $747.09
 
B56171G View Documents TCI America
B56171G
1g Each for $249.03
 
Specifications

Chemical Identifiers

117766-40-2
378.471
11783778
COC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)OC
C27H22O2
BYDIWLYAWBNCAQ-UHFFFAOYSA-N
9,9-bis(4-methoxyphenyl)fluorene

Specifications

White
5g
C27H22O2
COC1=CC=C(C=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC=C(C=C5)OC
378.471
378.47
≥98.0% (GC)
128°C
117766-40-2
BYDIWLYAWBNCAQ-UHFFFAOYSA-N
9,9-bis(4-methoxyphenyl)fluorene
11783778
Crystal-Powder at 20°C
9,9-Bis(4-methoxyphenyl)-9H-fluorene
Safety and Handling

Safety and Handling

TSCA : No