missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
9,9-Bis(4-amino-3-methylphenyl)fluorene 98.0+%, TCI America™
Supplier: TCI America B269325G
Specifications
9,9-Bis(4-amino-3-methylphenyl)fluorene | |
233°C | |
C27H24N2 | |
2,2′-Dimethyl-4,4′-(9-fluorenylidene)dianiline, 4,4′-(9-Fluorenylidene)di-o-toluidine | |
CC1=C(C=CC(=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC(=C(C=C5)N)C)N | |
376.503 | |
376.50 | |
Crystalline Powder |
107934-60-1 | |
White-Yellow | |
25 g | |
YRKVLGUIGNRYJX-UHFFFAOYSA-N | |
4-[9-(4-amino-3-methylphenyl)fluoren-9-yl]-2-methylaniline | |
21872150 | |
≥98.0% (T) |
Chemical Identifiers
107934-60-1 | |
376.503 | |
21872150 | |
CC1=C(C=CC(=C1)C2(C3=CC=CC=C3C4=CC=CC=C42)C5=CC(=C(C=C5)N)C)N |
C27H24N2 | |
YRKVLGUIGNRYJX-UHFFFAOYSA-N | |
4-[9-(4-amino-3-methylphenyl)fluoren-9-yl]-2-methylaniline |
Safety and Handling
TSCA : No