missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9'-Bifluorenyl 98.0+%, TCI America™
$162.17 - $529.40
Chemical Identifiers
| CAS | 1530-12-7 |
|---|---|
| Molecular Formula | C26H18 |
| Molecular Weight (g/mol) | 330.43 |
| MDL Number | MFCD00001132 |
| InChI Key | QXAIDADUIMVTPS-UHFFFAOYSA-N |
| PubChem CID | 137063 |
| IUPAC Name | 9-(9H-fluoren-9-yl)-9H-fluorene |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)C4C5=CC=CC=C5C6=CC=CC=C46 |
Chemical Identifiers
| 1530-12-7 | |
| 330.43 | |
| QXAIDADUIMVTPS-UHFFFAOYSA-N | |
| 9-(9H-fluoren-9-yl)-9H-fluorene |
| C26H18 | |
| MFCD00001132 | |
| 137063 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)C4C5=CC=CC=C5C6=CC=CC=C46 |
Specifications
| 1530-12-7 | |
| White | |
| MFCD00001132 | |
| 9,9′C-Bifluorene | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)C4C5=CC=CC=C5C6=CC=CC=C46 | |
| 330.43 | |
| 330.43 | |
| Crystalline Powder |
| 246°C | |
| C26H18 | |
| 1 g | |
| QXAIDADUIMVTPS-UHFFFAOYSA-N | |
| 9-(9H-fluoren-9-yl)-9H-fluorene | |
| 137063 | |
| ≥98.0% (HPLC) | |
| 9,9′-Bifluorenyl |
Safety and Handling
TSCA : No