missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(4-Iodophenyl)carbazole, TCI America™

$221.53 - $768.48

Chemical Identifiers

CAS 57103-15-8
Molecular Formula C18H12IN
Molecular Weight (g/mol) 369.205
MDL Number MFCD11617970
InChI Key CWZDMKAGSIDFBR-UHFFFAOYSA-N
PubChem CID 10832858
IUPAC Name 9-(4-iodophenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09611G View Documents TCI America
I09611G
1g Each for $221.53
 
I09615G View Documents TCI America
I09615G
5g Each for $768.48
 
Specifications

Chemical Identifiers

57103-15-8
369.205
CWZDMKAGSIDFBR-UHFFFAOYSA-N
9-(4-iodophenyl)carbazole
C18H12IN
MFCD11617970
10832858
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)I

Specifications

White-Yellow
1g
C18H12IN
CWZDMKAGSIDFBR-UHFFFAOYSA-N
9-(4-iodophenyl)carbazole
10832858
Crystal-Powder at 20°C
9-(4-Iodophenyl)carbazole
140°C
57103-15-8
MFCD11617970
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)I
369.205
369.21
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No