missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(4-Ethynylphenyl)carbazole, TCI America™

Manufacturer:  TCI America E10555G

 View more versions of this product

Catalog No. E10555G



Specifications

Specifications

9-(4-Ethynylphenyl)carbazole
106°C
262861-81-4
MFCD12024288
C#CC1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42
267.331
267.33
≥98.0% (HPLC,N)
White-Yellow
5g
C20H13N
JJIQKIUIIVFHAN-UHFFFAOYSA-N
9-(4-ethynylphenyl)carbazole
15606338
Crystal-Powder at 20°C

Chemical Identifiers

262861-81-4
267.331
JJIQKIUIIVFHAN-UHFFFAOYSA-N
9-(4-ethynylphenyl)carbazole
C20H13N
MFCD12024288
15606338
C#CC1=CC=C(C=C1)N2C3=CC=CC=C3C4=CC=CC=C42
Safety and Handling

Safety and Handling

TSCA : No

Documents