missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole, TCI America™

$235.28 - $702.80

Chemical Identifiers

CAS 601454-33-5
Molecular Formula C26H28BrN
Molecular Weight (g/mol) 434.421
MDL Number MFCD11617971
InChI Key LAHSPJHDQUQBTB-UHFFFAOYSA-N
PubChem CID 57604191
IUPAC Name 9-(4-bromophenyl)-3,6-ditert-butylcarbazole
SMILES CC(C)(C)C1=CC2=C(C=C1)N(C3=C2C=C(C=C3)C(C)(C)C)C4=CC=C(C=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B53131G View Documents TCI America
B53131G
1g Each for $235.28
 
B53135G View Documents TCI America
B53135G
5g Each for $702.80
 
Specifications

Chemical Identifiers

601454-33-5
434.421
LAHSPJHDQUQBTB-UHFFFAOYSA-N
9-(4-bromophenyl)-3,6-ditert-butylcarbazole
C26H28BrN
MFCD11617971
57604191
CC(C)(C)C1=CC2=C(C=C1)N(C3=C2C=C(C=C3)C(C)(C)C)C4=CC=C(C=C4)Br

Specifications

White
1g
C26H28BrN
LAHSPJHDQUQBTB-UHFFFAOYSA-N
9-(4-bromophenyl)-3,6-ditert-butylcarbazole
57604191
Crystal-Powder at 20°C
9-(4-Bromophenyl)-3,6-di-tert-butylcarbazole
156°C
601454-33-5
MFCD11617971
CC(C)(C)C1=CC2=C(C=C1)N(C3=C2C=C(C=C3)C(C)(C)C)C4=CC=C(C=C4)Br
434.421
434.42
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No