missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(4-Biphenylyl)carbazole, TCI America™

$158.89 - $554.59

Chemical Identifiers

CAS 6299-16-7
Molecular Formula C24H17N
Molecular Weight (g/mol) 319.407
MDL Number MFCD28147704
InChI Key DQMMBEPJQZXXGK-UHFFFAOYSA-N
PubChem CID 412897
IUPAC Name 9-(4-phenylphenyl)carbazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=CC=CC=C53
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B51281G View Documents TCI America
B51281G
1g Each for $158.89
 
B51285G View Documents TCI America
B51285G
5g Each for $554.59
 
Specifications

Chemical Identifiers

6299-16-7
319.407
DQMMBEPJQZXXGK-UHFFFAOYSA-N
9-(4-phenylphenyl)carbazole
C24H17N
MFCD28147704
412897
C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=CC=CC=C53

Specifications

White-Yellow
1g
C24H17N
DQMMBEPJQZXXGK-UHFFFAOYSA-N
9-(4-phenylphenyl)carbazole
412897
Crystal-Powder at 20°C
9-(4-Biphenylyl)carbazole
227°C
6299-16-7
MFCD28147704
C1=CC=C(C=C1)C2=CC=C(C=C2)N3C4=CC=CC=C4C5=CC=CC=C53
319.407
319.41
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No