missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(3-Bromophenyl)carbazole, TCI America™

$120.70 - $368.20

Chemical Identifiers

CAS 185112-61-2
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.205
MDL Number MFCD20486475
InChI Key ZKGHGKNHPPZALY-UHFFFAOYSA-N
PubChem CID 58690888
IUPAC Name 9-(3-bromophenyl)carbazole
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC(=CC=C4)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B44271G View Documents TCI America
B44271G
1g Each for $120.70
 
B44275G View Documents TCI America
B44275G
5g Each for $368.20
 
Specifications

Chemical Identifiers

185112-61-2
322.205
ZKGHGKNHPPZALY-UHFFFAOYSA-N
9-(3-bromophenyl)carbazole
C18H12BrN
MFCD20486475
58690888
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC(=CC=C4)Br

Specifications

White-Yellow
1g
C18H12BrN
ZKGHGKNHPPZALY-UHFFFAOYSA-N
9-(3-bromophenyl)carbazole
58690888
Crystal-Powder at 20°C
9-(3-Bromophenyl)carbazole
69°C
185112-61-2
MFCD20486475
C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC(=CC=C4)Br
322.205
322.21
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No