missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(2-Hydroxyethyl)adenine, TCI America™

$110.00 - $317.78

Chemical Identifiers

CAS 707-99-3
Molecular Formula C7H9N5O
Molecular Weight (g/mol) 179.183
MDL Number MFCD04220770
InChI Key VAQOTZQDXZDBJK-UHFFFAOYSA-N
Synonym 6-Amino-9-(2-hydroxyethyl)purine
PubChem CID 242652
IUPAC Name 2-(6-aminopurin-9-yl)ethanol
SMILES C1=NC2=C(C(=N1)N)N=CN2CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H12905G View Documents TCI America
H12905G
5g Each for $110.00
 
H129025G View Documents TCI America
H129025G
25g Each for $317.78
 
Specifications

Chemical Identifiers

707-99-3
179.183
VAQOTZQDXZDBJK-UHFFFAOYSA-N
242652
C1=NC2=C(C(=N1)N)N=CN2CCO
C7H9N5O
MFCD04220770
6-Amino-9-(2-hydroxyethyl)purine
2-(6-aminopurin-9-yl)ethanol

Specifications

White-Yellow
5g
C7H9N5O
6-Amino-9-(2-hydroxyethyl)purine
C1=NC2=C(C(=N1)N)N=CN2CCO
179.183
179.18
≥98.0% (HPLC,T)
242°C
707-99-3
MFCD04220770
VAQOTZQDXZDBJK-UHFFFAOYSA-N
2-(6-aminopurin-9-yl)ethanol
242652
Crystal-Powder at 20°C
9-(2-Hydroxyethyl)adenine
Safety and Handling

Safety and Handling

TSCA : No