missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9-(2-Ethylhexyl)carbazole, TCI America™

Manufacturer:  TCI America E09725G

 View more versions of this product

Catalog No. E09725G



Specifications

Specifications

9-(2-Ethylhexyl)carbazole
5g
C20H25N
SAXNQWFLHXTRDI-UHFFFAOYSA-N
9-(2-ethylhexyl)carbazole
16217868
Clear Liquid at 20°C
Yellow
187148-77-2
MFCD06798127
CCCCC(CC)CN1C2=CC=CC=C2C3=CC=CC=C31
279.427
279.43
≥97.0% (GC)

Chemical Identifiers

187148-77-2
279.427
SAXNQWFLHXTRDI-UHFFFAOYSA-N
9-(2-ethylhexyl)carbazole
C20H25N
MFCD06798127
16217868
CCCCC(CC)CN1C2=CC=CC=C2C3=CC=CC=C31
Safety and Handling

Safety and Handling

TSCA : No

Documents