missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9(10H)-Acridone, 99%

Manufacturer:  Alfa Aesar™ A1522214

 View more versions of this product

Catalog No. AAA1522214



Description

Description

Applications
9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

Solubility
Insoluble in benzene, chloroform, ether, water and ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

9(10H)-Acridone
25g
C13H9NO
7104
FZEYVTFCMJSGMP-UHFFFAOYSA-N
10H-acridin-9-one
2015
195.22
360°C to 362°C
578-95-0
MFCD00005019
acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2
195.221
CHEBI:50756
99%

Chemical Identifiers

578-95-0
195.221
FZEYVTFCMJSGMP-UHFFFAOYSA-N
2015
10H-acridin-9-one
C13H9NO
MFCD00005019
acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
CHEBI:50756
C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2
Safety and Handling

Safety and Handling

RTECSNumber : AR6976000

TSCA : TSCA

Documents