missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9,10-Phenanthrenequinone 99.0+%, TCI America™

Manufacturer:  TCI America P00805G

 View more versions of this product

Catalog No. P00805G



Specifications

Specifications

9,10-Phenanthrenequinone
210°C
84-11-7
MFCD00001163
9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O
208.216
CHEBI:37454
Crystal-Powder at 20°C
Undesignated
5g
C14H8O2
3077
YYVYAPXYZVYDHN-UHFFFAOYSA-N
phenanthrene-9,10-dione
6763
208.22
≥99.0% (GC)

Chemical Identifiers

84-11-7
208.216
YYVYAPXYZVYDHN-UHFFFAOYSA-N
6763
phenanthrene-9,10-dione
C14H8O2
MFCD00001163
9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
CHEBI:37454
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O
Safety and Handling

Safety and Handling

RTECSNumber : SF7875000

TSCA : Yes

Documents