missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,10-Diphenylanthracene, 99%

Manufacturer:  Alfa Aesar™ A1571414

 View more versions of this product

Catalog No. AAA1571414



Description

Description

Applications
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.

Solubility
Soluble in ethanol, chloroform, benzene and carbon disulfide. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

9,10-Diphenylanthracene
248°C to 254°C
1499-10-1
MFCD00001253
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)C5=CC=CC=C5
330.43
CHEBI:51676
99%
Odorless
25g
C26H18
1914010
FCNCGHJSNVOIKE-UHFFFAOYSA-N
9,10-diphenylanthracene
15159
330.43

Chemical Identifiers

1499-10-1
330.43
FCNCGHJSNVOIKE-UHFFFAOYSA-N
15159
9,10-diphenylanthracene
C26H18
MFCD00001253
anthracene, 9,10-diphenyl, 9,10-diphenyl anthracene, unii-51bq8iyq9u, 51bq8iyq9u, acmc-1brsc, 9, 9-diphenylanthracene, anthracene,10-diphenyl, 9,10-diphenylanthracene, 9,10-diphenylanthracene 1g, 9,10-diphenylanthracene, analytical standard
CHEBI:51676
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)C5=CC=CC=C5
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents