missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Dimethylanthracene 98.0+%, TCI America™
$182.68 - $920.84
Chemical Identifiers
| CAS | 781-43-1 |
|---|---|
| Molecular Formula | C16H14 |
| Molecular Weight (g/mol) | 206.29 |
| MDL Number | MFCD00001262 |
| InChI Key | JTGMTYWYUZDRBK-UHFFFAOYSA-N |
| Synonym | anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene |
| PubChem CID | 13076 |
| IUPAC Name | 9,10-dimethylanthracene |
| SMILES | CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12 |
Chemical Identifiers
| 781-43-1 | |
| 206.29 | |
| JTGMTYWYUZDRBK-UHFFFAOYSA-N | |
| 13076 | |
| CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12 |
| C16H14 | |
| MFCD00001262 | |
| anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene | |
| 9,10-dimethylanthracene |
Specifications
| 781-43-1 | |
| Yellow | |
| MFCD00001262 | |
| anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene | |
| CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12 | |
| 206.29 | |
| 206.29 | |
| Crystalline Powder |
| 182°C | |
| C16H14 | |
| 1 g | |
| JTGMTYWYUZDRBK-UHFFFAOYSA-N | |
| 9,10-dimethylanthracene | |
| 13076 | |
| ≥98.0% (GC) | |
| 9,10-Dimethylanthracene |
Safety and Handling
EINECSNumber : (4)-0640
RTECSNumber : CA9685000
TSCA : No