missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dimethylanthracene, 97%, Thermo Scientific™

Chemical Identifiers

CAS 781-43-1
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001262
InChI Key JTGMTYWYUZDRBK-UHFFFAOYSA-N
Synonym anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
PubChem CID 13076
IUPAC Name 9,10-dimethylanthracene
SMILES CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC115972500
SDS View Documents
Thermo Scientific Chemicals
115972500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC115970010
SDS View Documents
Thermo Scientific Chemicals
115970010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

781-43-1
206.29
JTGMTYWYUZDRBK-UHFFFAOYSA-N
13076
CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12
C16H14
MFCD00001262
anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
9,10-dimethylanthracene

Specifications

781-43-1
100.0
97%
C16H14
250mg
anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthracene
CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12
206.29
206.29
9, 10-Dimethylanthracene, 97%
96.0
Authentic
Glass bottle
MFCD00001262
05,II,593
JTGMTYWYUZDRBK-UHFFFAOYSA-N
9,10-dimethylanthracene
13076
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-308-4

RTECSNumber : CA9685000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only