missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,10-Bis(phenylethynyl)anthracene

As a photochromic material exhibiting chemiluminescence

Manufacturer:  Alfa Aesar™ 4331303

 View more versions of this product

Catalog No. AA4331303



Specifications

Specifications

9,10-Bis (phenylethynyl)anthracene
254°C to 255°C
10075-85-1
MFCD00012050
9,10-bis 2-phenylethynyl anthracene
C1=CC=C(C=C1)C#CC2=C3C=CC=CC3=C(C4=CC=CC=C42)C#CC5=CC=CC=C5
378.474
CHEBI:51675
Crystalline
Light Sensitive
1g
C30H18
1891432
ZHBOFZNNPZNWGB-UHFFFAOYSA-N
9,10-bis(2-phenylethynyl)anthracene
82338
378.47

Chemical Identifiers

10075-85-1
378.474
ZHBOFZNNPZNWGB-UHFFFAOYSA-N
82338
9,10-bis(2-phenylethynyl)anthracene
C30H18
MFCD00012050
9,10-bis 2-phenylethynyl anthracene
CHEBI:51675
C1=CC=C(C=C1)C#CC2=C3C=CC=CC3=C(C4=CC=CC=C42)C#CC5=CC=CC=C5
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents