missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 9,10-Bis(4-ethoxyphenyl)-2-chloroanthracene, 98%

Manufacturer:  Alfa Aesar™ H5336914

 View more versions of this product

Catalog No. AAH5336914



Specifications

Specifications

9,10-Bis(4-ethoxyphenyl)-2-chloroanthracene
25g
C30H25ClO2
2-chloro-9,10-bis 4-ethoxyphenyl anthracene, 9,10-bis 4-ethoxyphenyl-2-chloroanthracene, 9,10-bis-4-ethoxyphenyl-2-chloroanthracene, 9,10-bis 4-ethoxyphenyl-2-chloroanthracene
CCOC1=CC=C(C=C1)C2=C3C=CC(=CC3=C(C4=CC=CC=C42)C5=CC=C(C=C5)OCC)Cl
452.978
452.98
264°C to 265°C
135965-21-8
MFCD00190198
RFVZBUUPBPFZMH-UHFFFAOYSA-N
2-chloro-9,10-bis(4-ethoxyphenyl)anthracene
21715448
98%

Chemical Identifiers

135965-21-8
452.978
RFVZBUUPBPFZMH-UHFFFAOYSA-N
21715448
CCOC1=CC=C(C=C1)C2=C3C=CC(=CC3=C(C4=CC=CC=C42)C5=CC=C(C=C5)OCC)Cl
C30H25ClO2
MFCD00190198
2-chloro-9,10-bis 4-ethoxyphenyl anthracene, 9,10-bis 4-ethoxyphenyl-2-chloroanthracene, 9,10-bis-4-ethoxyphenyl-2-chloroanthracene, 9,10-bis 4-ethoxyphenyl-2-chloroanthracene
2-chloro-9,10-bis(4-ethoxyphenyl)anthracene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents