Learn More
9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate, 97% (dry wt.), water ca 4-6%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2496936
Description
9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate. It is used as a catalyst in production of alkaline pulping in the soda process. It is used as an in situ photochemical fluorescence probe.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate. It is used as a catalyst in production of alkaline pulping in the soda process. It is used as an in situ photochemical fluorescence probe.
Solubility
Soluble in hot water, cold water and insoluble in alcohol or ether.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate | |
>320°C | |
C14H7NaO5S | |
500 g | |
sodium anthraquinone-2-sulfonate, anthraquinone-2-sulfonic acid sodium salt, sodium 2-anthraquinonesulfonate, unii-511h72sdvx, sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, 2-anthracenesulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt, sodium 2-anthrachinonesulphonate, 2-anthraquinone sodium sulfonate, 2-anthraquinonesulfonic acid sodium salt, sodium-2-anthrachinonesulphonate | |
GGCZERPQGJTIQP-UHFFFAOYSA-M | |
sodium;9,10-dioxoanthracene-2-sulfonate | |
23661981 | |
97% |
153277-35-1 | |
(dry wt.), water ≈4 to 6% | |
MFCD00149068 | |
3580452 | |
Soluble in hot water,cold water and insoluble in alcohol or ether. | |
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)S(=O)(=O)[O-].[Na+] | |
310.255 | |
310.26 (Anhydrous) |
Chemical Identifiers
153277-35-1 | |
310.255 | |
GGCZERPQGJTIQP-UHFFFAOYSA-M | |
23661981 | |
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)S(=O)(=O)[O-].[Na+] |
C14H7NaO5S | |
MFCD00149068 | |
sodium anthraquinone-2-sulfonate, anthraquinone-2-sulfonic acid sodium salt, sodium 2-anthraquinonesulfonate, unii-511h72sdvx, sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, 2-anthracenesulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt, sodium 2-anthrachinonesulphonate, 2-anthraquinone sodium sulfonate, 2-anthraquinonesulfonic acid sodium salt, sodium-2-anthrachinonesulphonate | |
sodium;9,10-dioxoanthracene-2-sulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-009-5
RTECSNumber : CB1095550
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only