Thermo Scientific™ 8000 Series Silica Particle Size Standards
Provide greater durability and chemical stability when using solvents with these monodisperse silica microspheres used in instrument calibration and as spacers.
Supplier: Thermo Scientific™ 8050
Description
Provide greater durability and chemical stability when using solvents with Thermo Scientific™ 8000 Series Silica Particle Size Standards, monodisperse silica microspheres used in instrument calibration and as spacers. Standards are calibrated and certified with NIST™ traceable mean diameters and are suitable for a wide variety of particle measurement applications.
Features:
- Standards are designed for applications requiring monodisperse inorganic spheres.
- Opaque spheres provide more contrast than polymer particles in optical and electron beams; like glass, silica has a much higher density that polystyrene.
Specifications
Silica Particle Size Standard | |
0.49μm ± 0.02μm |