Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Undecenal, predominantly cis, 96%

$226.01 - $738.31

Chemical Identifiers

Chemical Name or Material 8-Undecenal
CAS 58296-81-4
Name Note predominantly cis
Molecular Formula C11H20O
Formula Weight 168.29
MDL Number MFCD00015577
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1756114

  SDS
alfa aesar™
A1756114
25g Each for $226.01

AAA1756122

  SDS
alfa aesar™
A1756122
100g Each for $738.31
Description & Specifications

Specifications

Formula Weight 168.29
TSCA TSCA
Assay 96%
MDL Number MFCD00015577
Molecular Formula C11H20O
Flash Point 93°C (199°F)
Boiling Point 95°C (4mmHg)
Melting Point 261° to 263°C
CAS 58296-81-4
Quantity 25g
Chemical Name or Material 8-Undecenal
Name Note predominantly cis