missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Phenyloctanoic acid, 97%

$79.58 - $327.86

Chemical Identifiers

CAS 26547-51-3
Molecular Formula C6H5(CH2)7CO2H
Formula Weight 220.31
MDL Number MFCD00037802
Beilstein 2050336
Chemical Name or Material 8-Phenyloctanoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1384303 SDS Alfa Aesar™
A1384303
1g Each for $79.58
AAA1384306 SDS Alfa Aesar™
A1384306
5g Each for $327.86
Specifications

Description

Applications
8-Phenyloctanoic acid is an n- phenylalkanoic acids which is an intermediate that support bacterial growth. Pseudomonas p utida disrupted in the gene that encodes the 3-ketoacyl-CoA thiolase (fadA), was cultured in a chemically defined medium containing 8-Phenyloctanoic acid as aromatic fatty acids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

26547-51-3
220.31
2050336
C6H5(CH2)7CO2H
MFCD00037802
8-Phenyloctanoic acid

Specifications

150° to 152°C (0.1mmHg)
1g
C6H5(CH2)7CO2H
MFCD00037802
8-Phenyloctanoic acid
27° to 29°C
26547-51-3
220.31
2050336
97%

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning