Please login to your online account to display your discounted pricing

(-)-8-phenylmenthol 97%, ACROS Organics™

$141.68 - $141.68

Chemical Identifiers

Chemical Name or Material (-)-8-Phenylmenthol
CAS 65253-04-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C16H24O
Formula Weight 232.37
MDL Number MFCD00010501
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC387950010

  SDS
thermo fisher scientific
AC387950010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC387952500

  SDS
thermo fisher scientific
AC387952500
Glass Bottle in Plastic Container 250mg Each for $141.68
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Viscous Liquid
Formula Weight 232.37
Quantity 1g
Color Clear colorless to light yellow
Density 1.0000g/mL
Specific Rotation Condition −24.00 (20.00°C c=2,C2H5OH)
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (-)-8-Phenylmenthol
Specific Rotation -22° to -26° (20°C, 589nm) (c=1, ethanol)
Assay Percent Range 97%
Molecular Formula C16H24O
MDL Number MFCD00010501
CAS 65253-04-5
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.5285 to 1.5315 (20°C, 589nm)
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.