Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Phenyl-1-octanol, 97%

$467.09 - $467.09

Chemical Identifiers

Chemical Name or Material 8-Phenyl-1-octanol
CAS 10472-97-6
Molecular Formula C6H5(CH2)8OH
Formula Weight 206.33
MDL Number MFCD00040918
Beilstein 2556146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1175806

  SDS
alfa aesar™
L1175806
5g Each for $467.09
Description & Specifications

Specifications

Formula Weight 206.33
Chemical Name or Material 8-Phenyl-1-octanol
MDL Number MFCD00040918
Assay 97%
Molecular Formula C6H5(CH2)8OH
Beilstein 2556146
Boiling Point 125° to 127°C (0.2mmHg)
Odor Odorless
Quantity 5g
CAS 10472-97-6

Applications
8-Phenyl-1-octanol is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.