missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Phenyl-1-octanol, 97%

Manufacturer:  Alfa Aesar™ L1175803

 View more versions of this product

Catalog No. AAL1175803



Description

Description

Applications
8-Phenyl-1-octanol is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

8-Phenyl-1-octanol
125°C to 127°C (0.2mmHg)
10472-97-6
MFCD00040918
8-phenyl-1-octanol, benzeneoctanol, 8-phenyl-n-octanol, benzene-octanol, 8-phenyloctanol, 8-phenyloctyl alcohol, pubchem20051, acmc-1c2oz, 8-phenyl-1-octanol gc
C1=CC=C(C=C1)CCCCCCCCO
206.329
206.33
Odorless
1g
C14H22O
2556146
MGIGATOGROSKNW-UHFFFAOYSA-N
8-phenyloctan-1-ol
517850
97%

Chemical Identifiers

10472-97-6
206.329
MGIGATOGROSKNW-UHFFFAOYSA-N
517850
C1=CC=C(C=C1)CCCCCCCCO
C14H22O
MFCD00040918
8-phenyl-1-octanol, benzeneoctanol, 8-phenyl-n-octanol, benzene-octanol, 8-phenyloctanol, 8-phenyloctyl alcohol, pubchem20051, acmc-1c2oz, 8-phenyl-1-octanol gc
8-phenyloctan-1-ol
Documents