missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Pentadecanamine 96.0+%, TCI America™
$404.79 - $1,214.38
Chemical Identifiers
CAS | 18618-64-9 |
---|---|
Molecular Formula | C15H33N |
Molecular Weight (g/mol) | 227.436 |
MDL Number | MFCD00087197 |
InChI Key | NTPSQPZXFVRULZ-UHFFFAOYSA-N |
Synonym | 1-Heptyloctylamine, 8-Aminopentadecane |
PubChem CID | 232907 |
IUPAC Name | pentadecan-8-amine |
SMILES | CCCCCCCC(CCCCCCC)N |
Chemical Identifiers
18618-64-9 | |
227.436 | |
NTPSQPZXFVRULZ-UHFFFAOYSA-N | |
232907 | |
CCCCCCCC(CCCCCCC)N |
C15H33N | |
MFCD00087197 | |
1-Heptyloctylamine, 8-Aminopentadecane | |
pentadecan-8-amine |
Specifications
18618-64-9 | |
C15H33N | |
1 mL | |
1-Heptyloctylamine, 8-Aminopentadecane | |
CCCCCCCC(CCCCCCC)N | |
227.436 | |
227.44 | |
Liquid |
117°C | |
MFCD00087197 | |
2735 | |
NTPSQPZXFVRULZ-UHFFFAOYSA-N | |
pentadecan-8-amine | |
232907 | |
≥96.0% (GC,T) | |
8-Pentadecanamine |
Safety and Handling
TSCA : No