missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-(p-Toluenesulfonylamino)quinoline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5538614

 View more versions of this product

Catalog No. AAH5538614



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

8-(p-Toluenesulfonylamino)quinoline
154°C to 158°C
C16H14N2O2S
25 g
8-tosylamino quinoline, 8-p-tosylaminoquinoline, benzenesulfonamide, 4-methyl-n-8-quinolinyl, 8-p-tolylsulfonamido quinoline, 8-p-tolylsulfonylamino quinoline, 8-p-toluenesulfonylamino quinoline, p-toluenesulfonamide, n-8-quinolyl, 4-methyl-n-quinolin-8-yl benzenesulfonamide, 8-4-tolylsulfonylamino quinoline, 4-methyl-n-8-quinolinylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)NC1=C2N=CC=CC2=CC=C1
298.36
298.36
Solid
10304-39-9
Brown to White
MFCD00168990
264159
ZSMKPYXVUIWTCT-UHFFFAOYSA-N
4-methyl-N-quinolin-8-ylbenzenesulfonamide
82533
97%

Chemical Identifiers

10304-39-9
298.36
ZSMKPYXVUIWTCT-UHFFFAOYSA-N
82533
C16H14N2O2S
MFCD00168990
8-tosylamino quinoline, 8-p-tosylaminoquinoline, benzenesulfonamide, 4-methyl-n-8-quinolinyl, 8-p-tolylsulfonamido quinoline, 8-p-tolylsulfonylamino quinoline, 8-p-toluenesulfonylamino quinoline, p-toluenesulfonamide, n-8-quinolyl, 4-methyl-n-quinolin-8-yl benzenesulfonamide, 8-4-tolylsulfonylamino quinoline, 4-methyl-n-8-quinolinylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)NC1=C2N=CC=CC2=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : DB2948850

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions