missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-(p-Toluenesulfonylamino)quinoline, 97%

Manufacturer:  Alfa Aesar™ H5538606

 View more versions of this product

Catalog No. AAH5538606



Specifications

Specifications

8-(p-Toluenesulfonylamino)quinoline
154°C to 158°C
10304-39-9
MFCD00168990
4-methyl-n-8-quinolinylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC3=C2N=CC=C3
298.36
298.36
97%
Brown to White
5g
C16H14N2O2S
264159
ZSMKPYXVUIWTCT-UHFFFAOYSA-N
4-methyl-N-quinolin-8-ylbenzenesulfonamide
82533
Solid

Chemical Identifiers

10304-39-9
298.36
ZSMKPYXVUIWTCT-UHFFFAOYSA-N
82533
CC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC3=C2N=CC=C3
C16H14N2O2S
MFCD00168990
4-methyl-n-8-quinolinylbenzenesulfonamide
4-methyl-N-quinolin-8-ylbenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : DB2948850

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents