missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Oxoadenosine, TCI America™

Manufacturer:  TCI America O04011G

 View more versions of this product

Catalog No. O04011G



Specifications

Specifications

8-Oxoadenosine
1g
C10H13N5O5
7,8-Dihydro-8-oxoadenosine, 8-Hydroxyadenosine
C1=NC2=C(C(=N1)N)NC(=O)N2C3C(C(C(O3)CO)O)O
283.244
283.24
≥98.0% (HPLC)
White-Yellow
29851-57-8
MFCD15145291
UEHOMUNTZPIBIL-DTUHVUQASA-N
6-amino-9-[(2R,3S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7H-purin-8-one
91972088
Crystal-Powder at 20°C

Chemical Identifiers

29851-57-8
283.244
UEHOMUNTZPIBIL-DTUHVUQASA-N
91972088
C1=NC2=C(C(=N1)N)NC(=O)N2C3C(C(C(O3)CO)O)O
C10H13N5O5
MFCD15145291
7,8-Dihydro-8-oxoadenosine, 8-Hydroxyadenosine
6-amino-9-[(2R,3S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7H-purin-8-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents