Please login to your online account to display your discounted pricing

8-Nitroquinoline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 8-Nitroquinoline
Name Note 98%
CAS 607-35-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H6N2O2
Formula Weight 174.16
MDL Number MFCD00006806
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC157080250

  SDS
fisher scientific acros
157080250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 174.16
Physical Form Crystalline Powder
Quantity 25g
Color Dark yellow
TSCA TSCA
EINECS Number 210-135-9
RTECS Number VC1925000
Melting Point 86° to 90°C
CAS Max % 100.0
Packaging Glass bottle
CAS Min % 97.5
Sulfated Ash 0.1% max.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Suspected of causing cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 8-Nitroquinoline
CAS 607-35-2
MDL Number MFCD00006806
Assay Percent Range 98%
Molecular Formula C9H6N2O2
Name Note 98%
Safety and Handling

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Suspected of causing cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.