Learn More
8-Methylquinoline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 303620250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
8-Methylquinoline | |
611-32-5 | |
100.0 | |
Yellow | |
245°C to 248°C | |
Authentic | |
Glass bottle | |
1.6140 to 1.6160 (20°C, 589nm) | |
25 g | |
quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888 | |
CC1=C2N=CC=CC2=CC=C1 | |
143.19 | |
CHEBI:48984 | |
97% |
97% | |
96.0 | |
-80°C | |
1.0500g/mL | |
104°C | |
96% min. (GC) | |
C10H9N | |
MFCD00006810 | |
1.05 | |
JRLTTZUODKEYDH-UHFFFAOYSA-N | |
8-methylquinoline | |
11910 | |
143.19 | |
Liquid |
Chemical Identifiers
611-32-5 | |
143.19 | |
JRLTTZUODKEYDH-UHFFFAOYSA-N | |
11910 | |
8-methylquinoline |
C10H9N | |
MFCD00006810 | |
quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888 | |
CHEBI:48984 | |
CC1=C2N=CC=CC2=CC=C1 |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 210-264-0
RTECSNumber : VC0562000
TSCA : TSCA
RUO – Research Use Only