missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride, 90%, Maybridge™

Chemical Identifiers

CAS 646477-45-4
Molecular Formula C8H18Cl2N2
Molecular Weight (g/mol) 213.146
MDL Number MFCD07366481
InChI Key KVYQKWPZQKGICY-UHFFFAOYSA-N
Synonym 3-alpha-aminotropane dihydrochloride, 3-amino-8-methyl-8-azabicyclo 3.2.1 octane dihydrochloride, 8-azabicyclo 3.2.1 octan-3-amine, 8-methyl-, dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-amine dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-aminedihydrochloride, 8-methyl-8-azabicyclo 3.2.1 oct-3-ylamine dihydrochloride, 8-methyl-8-azabicyclo 3.2.1 octan-3-amine dihydrochloride
PubChem CID 2775928
IUPAC Name 8-methyl-8-azabicyclo[3.2.1]octan-3-amine;dihydrochloride
SMILES CN1C2CCC1CC(C2)N.Cl.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB14320EA
SDS View Documents
Fisher Scientific Company
BTB14320EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

646477-45-4
213.146
KVYQKWPZQKGICY-UHFFFAOYSA-N
2775928
CN1C2CCC1CC(C2)N.Cl.Cl
C8H18Cl2N2
MFCD07366481
3-alpha-aminotropane dihydrochloride, 3-amino-8-methyl-8-azabicyclo 3.2.1 octane dihydrochloride, 8-azabicyclo 3.2.1 octan-3-amine, 8-methyl-, dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-amine dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-aminedihydrochloride, 8-methyl-8-azabicyclo 3.2.1 oct-3-ylamine dihydrochloride, 8-methyl-8-azabicyclo 3.2.1 octan-3-amine dihydrochloride
8-methyl-8-azabicyclo[3.2.1]octan-3-amine;dihydrochloride

Specifications

Amber Glass Bottle
646477-45-4
100.0
C8H18Cl2N2
3-alpha-aminotropane dihydrochloride, 3-amino-8-methyl-8-azabicyclo 3.2.1 octane dihydrochloride, 8-azabicyclo 3.2.1 octan-3-amine, 8-methyl-, dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-amine dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-aminedihydrochloride, 8-methyl-8-azabicyclo 3.2.1 oct-3-ylamine dihydrochloride, 8-methyl-8-azabicyclo 3.2.1 octan-3-amine dihydrochloride
CN1C2CCC1CC(C2)N.Cl.Cl
213.146
213.15
8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
10g
89.5
90%
MFCD07366481
KVYQKWPZQKGICY-UHFFFAOYSA-N
8-methyl-8-azabicyclo[3.2.1]octan-3-amine;dihydrochloride
2775928
90%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS